Merck
  • Novel neurotrophic effects of sphingosylphosphorylcholine in cerebellar granule neurons and in PC12 cells.
Novel neurotrophic effects of sphingosylphosphorylcholine in cerebellar granule neurons and in PC12 cells.
Biochemical and biophysical research communications (2007-10-16)
Naoko Konno, Ayako Nakamura, Yutaka Ikeno, So-hyun Cheon, Katsuhiko Kitamoto, Manabu Arioka
ABSTRACT
Sphingosylphosphorylcholine (SPC) is a choline-containing naturally occurring derivative of sphingolipid involved in various biological processes. Here we show that SPC displays neurotrophic effects in cerebellar granule neurons (CGNs) and in PC12 cells. When CGNs were cultured under non-depolarizing condition, they exhibited condensed and fragmented nuclei typical of apoptotic phenotype. SPC added to the culture medium rescued cells from undergoing apoptosis. The anti-apoptotic effect of SPC was dependent on the presence of extracellular Ca2+, suggesting that Ca2+ influx occurs upon SPC treatment. In PC12 cells, SPC displayed nerve growth factor-like neuritogenic effect which was sensitive to the presence of Ca2+ channel blocker and Ca2+ withdrawal from the medium. These results suggest that SPC plays novel neurotrophic effects in the nervous system.
Sigma-Aldrich
Sphingosylphosphorylcholine, ≥98%, powder