Skip to Content
Merck
All Photos(2)

Key Documents

HAWG050S6

Millipore

S-Pak® Membrane Filter

mixed cellulose esters (MCE), pore size 0.45 μm, filter diam. 50 mm, white gridded for general microbiological analysis

Synonym(s):

Membrane filters, Semi-permeable membranes, S-Pak Filters 0.45µm 50mm white gridded

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

material

gridded filter
mixed cellulose esters (MCE)
white filter

sterility

sterile; ethylene oxide treated

feature

easily wetted with water
hydrophilic
individually wrapped

manufacturer/tradename

S-Pak®

diam.

50 mm

filter diam.

50 mm

pore size

0.45 μm pore size

input

sample type soft drink
sample type sport drink
sample type beverage(s)
sample type beer
sample type wine
sample type food(s)
sample type cider
sample(s) (environmental)
sample type bottled water
sample type cosmetics

application(s)

bioburden testing
clinical
cosmetics
environmental
environmental
food and beverages
industrial qc

shipped in

ambient

General description

S-Pak Filters are made of Mixed cellulose esters (MCE) with 0.45 μm pore size, white gridded and 50mm diameter. These filters possess high recoveries and are more appropriate for general microbiological testing. S-Pak membranes and dispenser revolutionize filter handling for Membrane Filtration (MF) method testing by combining the benefits of ready-to-use, sterile, individually packed membranes with unique handling ease.

Application

The S-Pak system is suitable for testing drinking, bottled and mineral water according to international standards.
These filters find application in Clinical, Cosmetics, Environmental, Food and Beverage, Industrial QC and Pharmaceutical industry.
Microbial analysis of beer, bottled water, cider, sport drink and wine by membrane filtration method.

Packaging

Individually sealed, with blue separator paper, sterile

Legal Information

S-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service