Merck
  • Repeated exposure of rats to JP-4 vapor induces changes in neurobehavioral capacity and 5-HT/5-HIAA levels.
Repeated exposure of rats to JP-4 vapor induces changes in neurobehavioral capacity and 5-HT/5-HIAA levels.
Journal of toxicology and environmental health. Part A (1999-04-14)
A F Nordholm, J Rossi, G D Ritchie, S McInturf, M E Hulme, C McCool, L Narayanan, K L MacMahon, J Eggers, H F Leahy, R E Wolfe
ABSTRACT
Thirty-two Sprague-Dawley rats were exposed for 6 h/d for 14 consecutive days to JP-4 jet fuel vapor (2 mg/L) or room air control conditions. Following a 14- or 60-d recovery period, rats completed a battery of 8 tests selected from the Navy Neurobehavioral Toxicity Assessment Battery (NTAB) to evaluate changes in performance capacity. Exposure to JP-4 vapor resulted in significant changes in neurobehavioral capacity on several tests that varied as a function of the duration of the recovery period. Rats were evaluated for major neurotransmitter and metabolite levels in five brain regions and in the blood serum. Levels of 5-hydroxytryptamine (5-HT) and 5-hydroxyindoleacetic acid (5-HIAA) were shown to be significantly elevated in several brain regions as well as in the blood serum in the vapor-exposed groups. Results of the rat study are compared to previously reported neurobehavioral evaluations of European manufacturing personnel exposed chronically to jet fuel vapor.
Sigma-Aldrich
Kerosene, CP
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Kerosene, SAJ special grade, ≥90.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Kerosene, purum
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Kerosene, reagent grade, low odor