EP6001022410
Eppendorf® Upright Freezer Rack
Holds 8 x 102 mm/4 in boxes, (4 x 2) configuration, stainless steel, drawer
About This Item
Recommended Products
material
stainless steel
feature
(4 x 2) configuration
Holds 8 x 102 mm/4 in boxes
drawer
max. box number per rack: 8 (4 x 2)
max. box size [mm]: 136 x 136
manufacturer/tradename
Eppendorf® 6001022410 (Global)
technique(s)
cryopreservation: suitable
D × W × H
563 mm (22.2 in.) × 140 mm (5.5 in.) × 216 mm (8.5 in.)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service