Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS1997

Sigma-Aldrich

Anti-ST8SIA1 Antibody, clone HIC0-3C5

clone HIC0-3C5, from mouse

Synonym(s):

Alpha-N-acetylneuraminide alpha-2, 8-sialyltransferase, EC: 2.4.99.8, Alpha-2, 8-sialyltransferase 8A, Ganglioside GD3 synthase, Ganglioside GT3 synthase, Sialyltransferase 8A, SIAT8-A, Sialyltransferase St8Sia I, ST8SiaI

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$145.60
100 μG
$472.00

$145.60

List Price$182.00Save 20%

Estimated to ship onApril 17, 2025


Request a Bulk Order

Select a Size

Change View
25 μG
$145.60
100 μG
$472.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$145.60

List Price$182.00Save 20%

Estimated to ship onApril 17, 2025


Request a Bulk Order

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

HIC0-3C5, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... ST8SIA1(6489)

Related Categories

General description

Alpha-N-acetylneuraminide alpha-2,8-sialyltransferase (UniProt: Q92185; also known as EC: 2.4.99.8, Alpha-2,8-sialyltransferase 8A, Ganglioside GD3 synthase, Ganglioside GT3 synthase, Sialyltransferase 8A, SIAT8-A, Sialyltransferase St8Sia I, ST8SiaI) I encoded by the ST8SIA1 (also known as SIAT8, SIAT8A) gene (Gene ID: 6489) in human. SIAT8-A is a single-pass type II membrane protein that is strongly expressed in melanoma cell lines, adult and fetal brain, and to a lesser extent in adult and fetal lung. It is involved in the production of gangliosides GD3 and GT3 from GM3. It catalyzes the transfer of sialic acid from CMP-sialic acid to GM3 to produce GD3 and GT3 in a successive manner. It is also involved in the pathway protein glycosylation. SIAT8-A contains a cytoplasmic domain (aa 1-29); a transmembrane domain (aa 30-48); and a lumenal domain (aa 49-356). Two isoforms of SIAT8-A have been described that are produced by alternative splicing. (Ref.: Dorrell, C., et al. (2016). Nat. Commun. 7; 11756).

Specificity

Clone HIC0-3C5 specifically detects human Alpha-N-acetylneuraminide alpha-2,8-sialyltransferase (ST8SIA1).

Immunogen

Trypsin-dissociated human pancreatic islet cells with low levels of exocrine and ductal cells.

Application

Anti-ST8SIA1, clone HIC0-3C5, Cat. No. MABS1997, is a mouse monoclonal antibody that detects Alpha-N-acetylneuraminide alpha-2,8-sialyltransferase and has been tested for use in Flow Cytometry, Immunoflurescence, and Immunohistochemistry.
Flow Cytometry Analysis: A representative lot detected ST8SIA1 in Flow Cytometry applications (Dorrell, C., et. al. (2016). Nat Commun. 7:11756).

Immunofluorescence Analysis: A representative lot detected ST8SIA1 in Immunofluorescence applications (Dorrell, C., et. al. (2016). Nat Commun. 7:11756).
Research Category
Signaling

Quality

Evaluated by Immunohistochemistry in human pancreatic tissue sections.

Immunohistochemistry Analysis: A 1:250 dilution of this antibody detected ST8SIA1 in human pancreatic tissue sections.

Target description

40.52 kDa calculated.

Physical form

Format: Purified
Purified mouse monoclonal antibody IgM in PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service