biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
N3/26, monoclonal
mol wt
calculated mol wt 139.62 kDa
observed mol wt ~140 kDa
species reactivity
rat
packaging
antibody small pack of 100 μL
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable
isotype
IgG1κ
UniProt accession no.
shipped in
dry ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
rat ... Kcnt1(60444)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in rat brain tissue lysate.
Western Blotting Analysis: A 1:500 dilution of this antibody detected KCNT1 in rat brain tissue lysate.
Tested Applications
Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected KCNT1 in rat brain tissue sections.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service