8.14226
Trimethylsilylacetylene
for synthesis
Synonym(s):
Trimethylsilylacetylene, Ethinyltrimethylsilane
About This Item
Recommended Products
Quality Level
assay
≥98.0% (GC)
form
liquid
bp
52-53 °C
transition temp
flash point -34 °C
density
0.71 g/cm3 at 20 °C
storage temp.
2-30°C
InChI
1S/C5H10Si/c1-5-6(2,3)4/h1H,2-4H3
InChI key
CWMFRHBXRUITQE-UHFFFAOYSA-N
Analysis Note
Density (d 20 °C/ 4 °C): 0.709 - 0.711
Identity (IR): passes test
signalword
Danger
hcodes
Hazard Classifications
Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 1
flash_point_f
-29.2 °F
flash_point_c
-34 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service