MilliporeSigma
Mu and Delta Opioid Receptors Are Coexpressed and Functionally Interact in the Enteric Nervous System of the Mouse Colon.
Cellular and molecular gastroenterology and hepatology (2019-11-24)
Jesse J DiCello, Simona E Carbone, Ayame Saito, Pradeep Rajasekhar, Rhian A Ceredig, Vi Pham, Celine Valant, Arthur Christopoulos, Nicholas A Veldhuis, Meritxell Canals, Dominique Massotte, Daniel P Poole
RESUMEN
Functional interactions between the mu opioid receptor (MOR) and delta opioid receptor (DOR) represent a potential target for novel analgesics and may drive the effects of the clinically approved drug eluxadoline for the treatment of diarrhea-predominant irritable bowel syndrome. Although the enteric nervous system (ENS) is a likely site of action, the coexpression and potential interaction between MOR and DOR in the ENS are largely undefined. In the present study, we have characterized the distribution of MOR in the mouse ENS and examined MOR-DOR interactions by using pharmacologic and cell biology techniques. MOR and DOR expression was defined by using MORmCherry and MORmCherry-DOR-eGFP knockin mice. MOR-DOR interactions were assessed by using DOR-eGFP internalization assays and by pharmacologic analysis of neurogenic contractions of the colon. Although MOR was expressed by approximately half of all myenteric neurons, MOR-positive submucosal neurons were rarely observed. There was extensive overlap between MOR and DOR in both excitatory and inhibitory pathways involved in the coordination of intestinal motility. MOR and DOR can functionally interact, as shown through heterologous desensitization of MOR-dependent responses by DOR agonists. Functional evidence suggests that MOR and DOR may not exist as heteromers in the ENS. Pharmacologic studies show no evidence of cooperativity between MOR and DOR. DOR internalizes independently of MOR in myenteric neurons, and MOR-evoked contractions are unaffected by the sequestration of DOR. Collectively, these findings demonstrate that although MOR and DOR are coexpressed in the ENS and functionally interact, they are unlikely to exist as heteromers under physiological conditions.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anticuerpo anti-colina acetiltransferasa, Chemicon®, from goat
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
25 g
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 69.80
100 g
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 165.00
250 g
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 345.00
1 kg
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 983.00
5 kg
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 3,590.00
25 kg
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 11,860.00
25 kg
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 12,290.00
50 kg
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 19,880.00
Sigma-Aldrich
[D-Ala2, N-Me-Phe4, Gly5-ol]-Enkephalin acetate salt, ≥97% (HPLC)
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
25 g
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 69.80
100 g
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 165.00
250 g
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 345.00
1 kg
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 983.00
5 kg
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 3,590.00
25 kg
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 11,860.00
25 kg
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 12,290.00
50 kg
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 19,880.00
Sigma-Aldrich
CTOP, ≥97% (HPLC)
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
25 g
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 69.80
100 g
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 165.00
250 g
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 345.00
1 kg
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 983.00
5 kg
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 3,590.00
25 kg
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 11,860.00
25 kg
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 12,290.00
50 kg
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 19,880.00
Sigma-Aldrich
SNC80, ≥98% (HPLC), solid
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
25 g
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 69.80
100 g
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 165.00
250 g
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 345.00
1 kg
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 983.00
5 kg
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 3,590.00
25 kg
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 11,860.00
25 kg
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 12,290.00
50 kg
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 19,880.00
Sigma-Aldrich
Anti-Neuropeptide Y Antibody, serum, Chemicon®
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
25 g
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 69.80
100 g
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 165.00
250 g
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 345.00
1 kg
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 983.00
5 kg
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 3,590.00
25 kg
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 11,860.00
25 kg
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 12,290.00
50 kg
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
CAD 19,880.00