MilliporeSigma
MPEF-AAT-11DB-02
Zarbeco, LLC
Zarbeco Adjustable Articulated Arm Stand
with desktop base attachment
Sinónimos:
swing arm stand, stand for microscope
About This Item
PDF Icon
Shipped & Sold By Zarbeco, LLC
A Trusted Marketplace Seller
PDF Icon
Have a question?
General description
One knob adjustable articulated arm stand for the MiScope with desktop base attachment, arm is 11" long
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)