UMP3T-1
WPI
WPI Microinjection Syringe Pump
UltraMicroPump3 (one) and SMARTouch Controller
About This Item
Productos recomendados
General description
The touch-screen display on WPI's MICRO2T SMARTouch™ controller offers an intuitive, user-friendly interface. Our MICRO2T allows for simultaneous control of the UMP3 syringe pumps, and/or our NANOLITER2020 positive displacement pumps. Key features of the MICRO2T controller include a splash-proof touch screen, built-in memory configured for saving settings between use, and a dual mode motor drive. The UMP3 may be mounted to either a stereotaxic frame or a micro manipulator via its built-in mounting bar. An optional foot switch is available.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)