NF36BV
WPI
WPI NanoFil Needles
Beveled, 36G
About This Item
General description
The replaceable needles used with the NanoFil are available with either blunt or beveled tips. The blunt tip is used for injection into soft tissue and when a uniform solution distribution is needed. The beveled style is used for applications that involve the penetration of a tough tissue.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)