MilliporeSigma
V900627
Sigma-Aldrich
D-Aspartic acid
Vetec, reagent grade, 99%
Sinónimos:
(R)-(−)-Aminosuccinic acid, (R)-2-Aminosuccinic acid
About This Item
Fórmula lineal:
HO2CCH2CH(NH2)CO2H
Número de CAS:
Peso molecular:
133.10
Beilstein:
1723529
Número MDL:
Código UNSPSC:
12352209
ID de la sustancia en PubChem:
grado
reagent grade
Línea del producto
Vetec
Ensayo
99%
Formulario
powder
idoneidad de la reacción
reaction type: solution phase peptide synthesis
color
white to off-white
mp
>300 °C (lit.)
cadena SMILES
N[C@H](CC(O)=O)C(O)=O
InChI
1S/C4H7NO4/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H,6,7)(H,8,9)/t2-/m1/s1
Clave InChI
CKLJMWTZIZZHCS-UWTATZPHSA-N
Información legal
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Código de clase de almacenamiento
11 - Combustible Solids
Clase de riesgo para el agua (WGK)
WGK 1
Punto de inflamabilidad (°F)
Not applicable
Punto de inflamabilidad (°C)
Not applicable
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Edi Erwan et al.
Amino acids, 43(5), 1969-1976 (2012-04-03)
Intracerebroventricular (i.c.v.) administration of L-aspartate (L-Asp) attenuates stress responses in neonatal chicks, but the mechanism has not been clarified. In the present study, three behavioral experiments were carried out under socially isolated stressful conditions exacerbated by the use of corticotrophin-releasing
Nobutoshi Ota et al.
Amino acids, 43(5), 1873-1886 (2012-08-09)
D-Aspartate (D-Asp) is an endogenous amino acid in the central nervous and reproductive systems of vertebrates and invertebrates. High concentrations of D-Asp are found in distinct anatomical locations, suggesting that it has specific physiological roles in animals. Many of the
Yuichi Kaji et al.
The British journal of ophthalmology, 96(8), 1127-1131 (2012-06-15)
Gelatinous drop-like corneal dystrophy (GDLD), also known as familial subepithelial corneal amyloidosis, is an autosomal recessive disorder that causes progressive corneal opacity due to accumulation of amyloid fibrils in the corneal stroma. Genetic analyses have revealed that a mutation in
Antimo D'Aniello
Brain research reviews, 53(2), 215-234 (2006-11-23)
D-Aspartic acid (d-Asp), an endogenous amino acid present in vertebrates and invertebrates, plays an important role in the neuroendocrine system, as well as in the development of the nervous system. During the embryonic stage of birds and the early postnatal
Akifumi Oda et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 879(29), 3337-3343 (2011-08-30)
Molecular dynamics simulations of amyloid β(1-42) containing D-aspartic acid residues were performed using several continuous solvent models to investigate the usefulness of simulation methods for D-amino acid-containing proteins and peptides. Normal molecular dynamics simulations and replica exchange molecular dynamics simulations