Key Documents
RPC1000C
TriForest
TriForest Autoclavable Polycarbonate Boston Round Media Bottle
1000mL, 48 per case
Sinónimos:
Autoclavable Bottles, Polycarbonate Bottles, Boston Round Media Bottles
About This Item
PDF Icon
Shipped & Sold By TriForest
A Trusted Marketplace Seller
PDF Icon
Have a question?
feature
with cap
manufacturer/tradename
TriForest Enterprises
General description
TriForest polycarbonate (PC) labware is a safer alternative to glass. These bottles are designed to be utilized in most shaking incubators. Ideal for steam sterilization of all biologicals, liquids, reagents, media, buffers and aqueous solutions.
  • Made with virgin polycarbonate resin, durable, chemically inert, non-leaching, optically clear
  • Molded seal ring inside cap fits tightly against bottle neck
  • No fluid entrapment and back flow contamination
  • Fits most shaking incubators
  • Ideal for steam sterilization
  • Manufactured in a Class 10K cleanroom
  • Sold by the case - 48 bottles per case