PGC2000M-C
TriForest
TriForest PETG Square Media / Packaging Bottle
2000mL, 12 per case
Sinónimos:
PETG Bottles, Media Packaging Bottles
About This Item
Productos recomendados
feature
with cap
manufacturer/tradename
TriForest Enterprises
General description
TriForest PETG Media / Packaging bottles are ergonomic, clear, and leakproof. Designed with automation in mind, these bottles are optimal for all filling, capping, and packaging operations. They are heavy duty and break-resistant, ideal for storage of valuable sera.
- Thick walled and durable; shatter resistant
- Excellent gas barrier properties; reduced permeability to CO2/O2
- Molded-in graduations for approximate volume reference
- Compliant with USP Class VI and FDA guidelines
- Bottle and cap material are Certified Animal Derived Component Free
- Manufactured in a Class 10K cleanroom
- Sold by the case - 12 bottles per case
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)