Key Documents
55216
Supelco
Frit Removing Tool for Supelco® Analytical Columns
About This Item
UNSPSC Code:
41115700
Application
Extrae fritas de columnas analíticas de Supelco de 2,1mm a 4,6mm
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)