MilliporeSigma
13982-U
Supelco
Packed GC Column Set
suitable for 2261 per GPA (5-column set)
About This Item
Código UNSPSC:
41115710
eCl@ss:
32110502
NACRES:
SB.54
Agency
suitable for GPA 2261 (5-column set)
técnicas
gas chromatography (GC): suitable
aplicaciones
pharmaceutical (small molecule)
tipo de columna
packed GC
Descripción general
Gas chromatography column are of basically two types packed column and open tubular column. In packed column the carrier gas must percolate through its porous bed. The shape and physical form of the packed column depends upon the dimensions of the chromatograph oven, the nature of the sample, the sample load and the column inlet pressure.
This column set is specifically designed for a specific column switching application. Column manufacturing procedurs have been optimized resulting in column sets that provide highly reproducible analyses.
Aplicación
Packed GC column with flame ionization detector (FID) may be used for the qualitative and quantitative analysis of gasoline and associated contaminated groundwater. It may also be used with electron capture detector (ECD) for the qualitative and quantitative analysis of halogenated compounds present in gasoline.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Introduction to Analytical Gas Chromatography.
Scott RPW.
Science, 101-104 (1997)
Capillary and packed-column gas chromatography of gasoline hydrocarbons and EDB.
Bruell CJ and Hoag GE.
Science, 5-7 (1984)