Saltar al contenido
MilliporeSigma
Todas las fotos(12)

Documentos clave

ZRB2330

Sigma-Aldrich

Anti-CD5 Antibody, clone 1C19 Antibody, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sinónimos:

T-cell surface glycoprotein CD5;Lymphocyte antigen T1/Leu-1

Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C19, recombinant monoclonal

description

1C19 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 54.58 kDa
observed mol wt ~55 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1C19 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Human CD5. It targets an epitope within 19 amino acids from the N-terminal, extracellular domain.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from the N-terminal, extracellular domain of human CD5.

Application

Quality Control Testing

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CD5 in Jurkat cell lysate.

Tested applications

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected CD5 in human tonsil tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected CD5 in Jurkat cells..

Affinity Binding Assay: A representative lot of this antibody bound CD5 peptide with a KD of 1.0 x 10-12 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

T-cell surface glycoprotein CD5 (UniProt: P06127; also known as Lymphocyte antigen T1/Leu-1, CD5) is encoded by the CD5 (also known as LEU1) gene (Gene ID: 921) in human. CD5 is a single-pass type I membrane protein that is synthesized with a signal peptide (aa 1-24), which is subsequently cleaved off to produce the mature form that contains an extracellular domain (aa 25-372), a transmembrane domain (aa 373-402), and a cytoplasmic domain (aa 403-495). The extracellular region contains three scavenger receptor cysteine rich (SRCR) domains. CD5 is expressed on the surface of all T cells bearing the β heterodimer T cell receptor (TCR) but is absent or present at lower density on the T cell subset. It is involved in the regulation of T cell function and proliferation. It is also detected on a subset of B cells, specifically the B1 cells. However, B cells usually express two-to-five-fold fewer CD5 molecules per cell than T cells. In B cells, CD5 promotes constitutive activation of pERK1/2 in a calcium-dependent manner and involves the non-selective TRPC1 channel. In the C-terminal region, CD5 contains four tyrosine residues that can be phosphorylated by Lyn kinase to create binding sies for SHP-1 (PTPN6). CD5 is shown to be constitutively associated with SHP-1 and their interaction increases upon TCR stimulation. In mature, peripheral T cells, CD5 is present in lipid rafts of the T cell surface, where it promotes CD3 redistribution into rafts, which upregulates the activation of ZAP-70, linker for activation of T cells (LAT), and Ca2+ influx. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Voisinne, G., et al. (2018). Front. Immunol. 9; 2900; Garaud, S., et al. (2018). Cell. Mol. Immunol. 15(2); 158-170).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

¿No encuentra el producto adecuado?  

Pruebe nuestro Herramienta de selección de productos.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Elija entre una de las versiones más recientes:

Certificados de análisis (COA)

Lot/Batch Number

¿No ve la versión correcta?

Si necesita una versión concreta, puede buscar un certificado específico por el número de lote.

¿Ya tiene este producto?

Encuentre la documentación para los productos que ha comprado recientemente en la Biblioteca de documentos.

Visite la Librería de documentos

Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.

Póngase en contacto con el Servicio técnico