Documentos clave
Z742101
Cell Strainer
yellow, Individually Wrapped, sterile; γ-irradiated, pack of 50 ea
About This Item
UNSPSC Code:
41104932
NACRES:
NB.22
material
Nylon
yellow
sterility
sterile; γ-irradiated
packaging
pack of 50 ea
pkg of (individually wrapped)
manufacturer/tradename
MIDSCI SG-100ICS
pore size
100 μm
General description
Cell Strainers from MIDSCI can be used for many applications including isolating cells and removing clumps or debris. Cell Strainers from MIDSCI are compatible with standard 50mL centrifuge tubes. The exclusive Pestle design improves manipulation of material in the strainer during operation.
  • Polypropylene housing with nylon mesh filter eliminates the need to fabricate filters from gauze, paper, nylon mesh, or other materials
  • Elongated lip on housing extends beyond edge of centrifuge tube enabling aseptic handling with forceps
  • Extended gripping tab on housing for easy handling
  • Available in three porosities: 40, 70, 100 um
  • Color coded by mesh porosity for easy identification
  • Packaged sterile (gamma irradiated) in individually wrapped peel-to-open paper/plastic wrapper or bulk packed in bags
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
C C F Amaral et al.
International endodontic journal, 51(12), 1434-1445 (2018-05-16)
To evaluate the effects of the [NaF 12 g L-1  + NaCl 1 g L-1 ] solution used in the electrochemical dissolution process of fractured endodontic files, as well as its NiTi-containing product, on dentine hardness, topography and human fibroblast viability. Sixty single-rooted human teeth