Z376957
Gel Handler™ gel supports
sheet W × L × thickness 8 1/2 in. × 11 in. × 5 mil (0.125 mm)
About This Item
Código UNSPSC:
41105300
NACRES:
NB.51
Productos recomendados
envase
pkg of 10 sheets
lámina An × L × grosor
8 1/2 in. × 11 in. × 5 mil (0.125 mm)
Descripción general
UV-transparent polyolefin Gel-Handler sheets easily slide under a gel resting in solution for transport to bench or transilluminator. The sheet is sturdy, but it conforms to the shape of the gel, reducing the trapping of air bubbles. Sheets are chemically inert, autoclavable and reusable.
May be cut to size with scissors.
May be cut to size with scissors.
Información legal
Gel Handler is a trademark of C.B.S. Scientific Co.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)