Documentos clave
Z336815
BRAND® Imhoff cone rack
PMMA
Seleccione un Tamaño
1 EA
$190.00
$190.00
0 left in stock
Seleccione un Tamaño
Cambiar Vistas
1 EA
$190.00
About This Item
Código UNSPSC:
41122800
NACRES:
NB.35
$190.00
0 left in stock
Materiales
PMMA
Características
Holds 2 x Imhoff cones
fabricante / nombre comercial
BRAND 388060
BRAND
L × An × Al
300 mm × 130 mm × 400 mm
Descripción general
The BRAND® Imhoff Cone Rack is designed to securely hold two Imhoff sedimentation cones made of plastic, providing a practical solution for sedimentation processes. Constructed from a combination of PMMA and PP, this rack offers durability and stability while remaining lightweight. Its compact design makes it easy to carry, even when the cones are filled, facilitating convenient transport and handling in laboratory settings. This essential accessory enhances the efficiency of sedimentation experiments, ensuring that your cones are securely supported and easily accessible.
Otras notas
BR388060 (India)
Información legal
BRAND is a registered trademark of BRAND GMBH + CO KG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)