Documentos clave
P3733
Hamilton® Softgrip® fixed-volume pipette
volume 10 μL
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
descripción
At 100% volume (Accuracy within ±0.80%, Precision within ±0.40%)
Características
electronic: no
single-channel
envase
pack of 1 ea
fabricante / nombre comercial
Hamilton 55019-03
volumen
10 μL
color
purple
Descripción general
Same ergonomic features as the Hamilton Softgrip adjustable volume pipette.
  • Less discomfort in the hand and wrist as compared to other pipettes during prolonged use
  • Multi-point color coding for easy volume identification
Every SoftGrip pipette is calibrated at Hamilton with standards traceable to NIST. A certificate of calibration documenting accuracy and precision is provided in the package along with non-wettable recalibration labels for field service and recalibration.
Información legal
Hamilton is a registered trademark of Hamilton Co.
Softgrip is a registered trademark of Hamilton Co.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)