Documentos clave
I1768
Sigma-Aldrich
In Situ Hybridization: A Practical Approach
About This Item
ISBN-10:
0-19-963327-4
ISBN-13:
978-0-19-963327-2
Código UNSPSC:
55101509
info de publicaciones
D.G. Wilkinson, ed., IRL Press at Oxford University Press, Oxford, England, 1992, 163 pp., soft cover
fabricante / nombre comercial
Oxford University Press
Descripción general
Step-by-step protocols are presented for the preparation of cloned DNA, RNA and oligonucleotide probes, with both radioactive and non-radioactive labels, and their hybridization to mRNA, viral DNA and chromosomal DNA.
Índice
1. The Theory and Practice of In Situ Hybridization, D.G. Wilkinson
2. In Situ Hybridization to Cellular RNA with Radiolabelled RNA Probes, L.M. Angerer and R.C. Angerer
3. In Situ Hybridization with Oligodeoxyribonucleotide Probes, W. Scott Young, III
4. In Situ Hybridization with Biotinylated Probes, P. Emson and M.J. Gait
5. Whole Mount In Situ Hybridization in Drosophila, D. Tautz, M. Hulskamp, and R.J. Sommer
6. Whole Mount In Situ Hybridization of Vertebrate Embryos, D.G. Wilkinson
7. Simultaneous Detection of Cellular RNA and Proteins, M. Brahic and S. Ozden
8. In Situ Hybridization at the Electron Microscope Level, M. Binder
9. Detection of Virus in Infected Human Tissue by In Situ Hybridization, F.A. Lewis and M. Wells
10. In Situ Hybridization to Chromosomes with Biotinylated Probes, E. Viegas-Pequignot
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)