Documentos clave
G6897
Gradient-forming devices
total capacity 50 mL
About This Item
Código UNSPSC:
41100000
capacidad total
50 mL
Descripción general
Constructed of two equal-diameter heavy-walled acrylic cylinders secured to an acrylic base. PTFE stopcock between cylinders is closed for filling, then opened. Exit port from mixing chamber is controlled by addition of hose and pinch clamp. Geometry permits use of magnetic stirring bar in mixing chamber.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)