EP5895151000
Eppendorf® Rotor Lid
for FA-6x50
About This Item
Productos recomendados
feature
for FA-6x50
manufacturer/tradename
5895151000
Eppendorf® 5895151000
Eppendorf®
technique(s)
centrifugation: suitable
compatibility
for use with 5910 R
for use with 5920 R
Categorías relacionadas
Legal Information
Eppendorf is a registered trademark of Eppendorf AG