Documentos
EP1368
Sigma-Aldrich
Sigma-Aldrich® CVS10 comb
10 sample, thickness 2.0 mm
About This Item
UNSPSC Code:
41100000
NACRES:
NC.01
thickness
2.0 mm
well volume
80 μL
General description
Comb Prep number indicates the number of preparatory wells. Marker number indicates the number of small volume wells intended for molecular weight markers. MC indicates multichannel pipette compatible.
Legal Information
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany