Documentos clave
EP022822136
Rotor for Eppendorf® Vacufuge® Plus Concentrator
F-45-16-20, non aerosol tight, (16 x 6.5/10.0 mL flat-bottom tubes)
About This Item
UNSPSC Code:
41103909
feature
to hold 16 x 20x42 - 55 mm tubes
to hold 16 x 6.5/10 mL flat-bottom tubes
fixed angle rotor
non aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022822136 (NA)
technique(s)
centrifugation: suitable
capacity
(16 x 6.5/10.0 mL flat-bottom tubes)
compatibility
for use with centrifuge models Vacufuge Plus Concentrator
Categorías relacionadas
General description
Selection of rotors for the Eppendorf Vacufuge Plus.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Vacufuge is a registered trademark of Eppendorf AG