MRC-5 SV1 TG2
85042502, human lung (fetal), Fibroblast
About This Item
Productos recomendados
product name
MRC-5 SV1 TG2, 85042502
biological source
human lung (fetal)
growth mode
Adherent
karyotype
Modal no. = 60-80 hperdiploid
morphology
Fibroblast
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human foetal lung, SV40 transformed
Cell Line Description
A clonal derivative of an SV40 transformed permanent line of MRC-5. Selection was made using X-ray induction in the presence of 2μg/ml thioguanine. The cells are HAT sensitive.
DNA Profile
STR-PCR Data: Amelogenin: X,Y
CSF1PO: 11,12
D13S317: 11,14
D16S539: 11
D5S818: 11,12
D7S820: 10
THO1: 8
TPOX: 8
vWA: 15
CSF1PO: 11,12
D13S317: 11,14
D16S539: 11
D5S818: 11,12
D7S820: 10
THO1: 8
TPOX: 8
vWA: 15
Culture Medium
EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:5 to 1:10 i.e. seeding at 1-2x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.