Saltar al contenido
MilliporeSigma
Todas las fotos(1)

Documentos clave

VP229

5092804, human breast, Epithelial

Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización


About This Item

UNSPSC Code:
41106514

product name

VP229, 05092804

biological source

human breast

growth mode

Adherent

karyotype

Mean no. 74, modal no. 66, range 55-134

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Breast Cancer

Cell Line Description

Derived from a breast cancer biopsy removed from a 47 year-old female. The histology of the breast cancer biopsy was defined as Ductal Grade III, tumour cells did not stain positive for oestrogen receptors. The VP229 cell line was derived from the same individual as the cell line VP267. VP229 was derived from the primary tumour and VP267 was derived 14 months later from a local recurrence of the tumour. The individual was recorded to live for 2 years after surgery performed to remove the primary tumour. This cell line grows slowly and is relatively difficult to culture; please read the sub-culture routine information closely.

Culture Medium

MEBM (Clonetics, CC-3151) + MEGM SingleQuots (Clonetics, CC-4136) + 2mM Glutamine + 2% Foetal Bovine Serum (FBS). Fresh EGF (final concentration of 10ng/ml) should be added to the medium at each subculture.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 i.e. seeding at 2-5x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C. The VP series of breast cancer cell lines are very slow growing and relatively difficult to grow. Fresh EGF (final concentration 10ng/ml) s

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificados de análisis (COA)

Busque Certificados de análisis (COA) introduciendo el número de lote del producto. Los números de lote se encuentran en la etiqueta del producto después de las palabras «Lot» o «Batch»

¿Ya tiene este producto?

Encuentre la documentación para los productos que ha comprado recientemente en la Biblioteca de documentos.

Visite la Librería de documentos

Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.

Póngase en contacto con el Servicio técnico