MilliporeSigma
211885
Sigma-Aldrich
Darrow Red
certified by the Biological Stain Commission, Dye content ≥65 %
About This Item
Fórmula empírica (notación de Hill):
C18H13N3O2
Número de CAS:
Peso molecular:
303.31
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
grade
certified by the Biological Stain Commission
form
powder
composition
Dye content, ≥65%
λmax
502 nm
ε (extinction coefficient)
13100 at 255 nm in 50% ethanol
9300 at 498 nm in 50% ethanol
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
Cl.CC(=O)Nc1ccc2N=C3C(Oc2c1)=CC(=N)c4ccccc34
InChI
1S/C18H13N3O2.ClH/c1-10(22)20-11-6-7-15-16(8-11)23-17-9-14(19)12-4-2-3-5-13(12)18(17)21-15;/h2-9,19H,1H3,(H,20,22);1H
InChI key
CJHKAKZHZRBAGI-UHFFFAOYSA-N
Suitability
Certified for the staining of neuron cell bodies in paraffin or frozen sections of formalin fixed brain or spinal cord.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificados de análisis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
¿Ya tiene este producto?
Visite la Librería de documentos
Kelly Del Tredici et al.
Journal of neuropathology and experimental neurology, 79(4), 378-392 (2020-02-06)
Although Gaucher disease can be accompanied by Lewy pathology (LP) and extrapyramidal symptoms, it is unknown if LP exists in Fabry disease (FD), another progressive multisystem lysosomal storage disorder. We aimed to elucidate the distribution patterns of FD-related inclusions and
Alexis Fenyi et al.
Cells, 10(1) (2021-01-16)
We investigated α-synuclein's (αSyn) seeding activity in tissue from the brain and enteric nervous system. Specifically, we assessed the seeding propensity of pathogenic αSyn in formalin-fixed tissue from the gastric cardia and five brain regions of 29 individuals (12 Parkinson's
Analysis and testing of biological stains--the Biological Stain Commission Procedures.
D P Penney et al.
Biotechnic & histochemistry : official publication of the Biological Stain Commission, 77(5-6), 237-275 (2003-02-05)