OCT-15B
Oxford Lab Products
Oxford Lab BenchMate Centrifuge Tubes
15mL, Sterile, 25 tubes x 20 packs, 500 tubes / case
About This Item
Productos recomendados
General description
Oxford Lab Centrifuge Tubes/ Conical Tubes
Oxford Lab Products Centrifuge Tubes (Conical Tubes) are designed to be secure, durable, and convenient. The material, engineering, and construction ensure that users can easily access their samples and store them safely.
Features
- The lid and seal design creates a leak-proof closure to prevent accidental sample loss.
- Clear and large graduations on the tube body and transparent surface.
- Large writing surface for easy marking and labeling.
- Oxford Centrifuge Tubes can endure temperatures ranging from -80 to 121°C
- Oxford Centrifuge Tubes are compatible with all conventional and high-capacity centrifuge rotors.
- Oxford Centrifuge Tubes are available in 15mL and 50mL sizes.
Properties
- Automatic human touch-free manufacturing manufactured in a 100k cleanroom environment
- Swiss-Made Diamond-Finished Mold creates a flawless super smooth internal surface
- Highly Durable Tubes can withstand up to 200 x g
- DNase, RNase, and Endotoxin Free every batch is tested for these by a GLP certified lab
- Super leak-proof seal design protects samples even in highly stressful conditions.
- High-Quality Materials made from US FDA Approved Virgin Polypropylene.
- Clear Markings and Labeling tamper-proof graduation markings and Labeling Area
Oxford Lab Products Centrifuge Tubes provide the reliability, security, and convenience expected from centrifuge tubes. These centrifuge tubes have everything that users need for their experiments: the lids offer secure seals, the plastic is durable, and the markings are accurate. The design of these tubes means that they can survive heat, cold, and up to 17,000 x g. We expect users to be comfortable using Oxford Lab Products Conical Tubes throughout all their experiments as they see fit.
We have taken extra care to use only the best materials—Oxford Lab Products. Conical tubes are made with FDA-approved resin. We use this plastic specifically to allow for extra clarity and durability. Our QC and manufacturing methods further ensure that customers receive only clean, contaminant-free, reliable, and sterile tubes that can be trusted to hold their samples without concern. Our manufacturing is fully automatic, human touch-free, and certified for compliance with quality systems. Try out our tubes today and work with peace of mind knowing that your samples are safe!