LTR-1000-LR
Oxford Lab Products
Oxford Lab Pipette Tips
1000uL LTS Compatible, Low Retention, Reload Stack, 96 Tips/Reload, 5 Reloads/Stack, 10 Stacks/Case.
About This Item
Productos recomendados
fabricante / nombre comercial
Oxford Lab Products
Descripción general
Oxford Lab BenchMate LTR Pipette Tips
Oxford BenchMate LTR Pipette Tips are compatible with Rainin LTS style pipettes. LTR Pipette tips offer the performance and quality you want without the harsh price tag you expect.
Features
- LTR Pipette Tips are manufactured using a swiss-made mold. The mold is diamond-finished to ensure that the features are fully and carefully constructed.
- BenchMate LTR Pipette Tips are designed to have an easy and secure attachment. The tip seal requires minimal force but securely attaches to the pipette.
- Oxford Lab Products requires strict Quality Control and Assurance procedures to ensure that tips are manufactured to a high standard.
- BenchMate LTR PIpette Tips are avaialble as sterile, low-retention, filtered tips to offer optimal pipetting performance.
- LTR Pipette Tips are designed to fit LTS or XLS Rainin Pipettes
- The tip design includes an ultra fine point that guarantees additional accuracy, precision, and consistency.
- Free of RNase, DNase, Cytotoxins, PCR Inhbitors, Endotoxins, and Pyrogens
- Internal surface is coated with a contaminant-free hydrophobic resin to ensure optimal liquid delivery.
Properties
- Swiss-Made Diamond Finished Mold flawless, super-smooth internal surface, with minimal retention.
- High-Quality Materials LTR Tips are made from US FDA approved Virgin Polypropylene
- Graduation Marked Tips LTR Pipette Tips are marked with Graduation markings as an additional accuracy/precision aid.
- Free From Softening Agents avoid potential contamination found in low-quality pipette tips.
- LTS Compatible can be used with Rainin LTS™ Pipettes
- Light attachment Force can be securely attached with minimal attachment force.
- Most Affordable on the Market BenchMate LTR Pipettes are the most affordable LTS™ compatible pipette tip
Filter Tips
- LTR Filtered PIpete tips are equipped with specially developed PE filters to prevent contamination of samples and pipettes.
- Filter placement is optimized to protect sample and pipette throughout prolonged use.
Non-Filter Tips
- Available in Low-Retention and/or sterile formats
Reloads/Refill Stack
- Available for Non-Filter Tips
- Compatible with Oxford BenchMate LTR Pipette Tips Racks
- Case includes 10 Towers of Reload Wafers
- Each Reload Stack includes 10 Reload Wafers, except 1000uL configuration which has 5 Reload Wafers.
Our LTR tips are available for single-channel pipettes as well as 8, 10, or 12 multi-channel LTS/XLS pipettes. Oxford Lab Products LTR tips are internally coated with hydrophobic, harm-free, low-retention resins to minimize sample retention. These tips are free of heavy metals and are free of RNase, DNase, cytotoxic materials, PCR inhibitors, endotoxins, and pyrogens to avoid sample contamination. The design of Oxford Lab Products LTR Tips also includes a fine-point end that yields additional accuracy, precision, and consistency. All our LTR tips are quality tested by our in-house lab technicians. Our QC protocols ensure that all Oxford Lab Products LTR tips are manufactured to meet strict regulations that enable high-performance pipetting.
Información legal
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documentos section.
Si necesita más asistencia, póngase en contacto con Atención al cliente