MilliporeSigma
ADR-30
Oxford Lab Products
Oxford Lab AccuTop Bottle Top Dispenser
2.5-30 ml, With Re-Circulation
About This Item
PDF Icon
Shipped & Sold By Oxford Lab Products
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
Oxford Lab Products
General description
Oxford Lab AccuTop Bottle Top Dispensers
The AccuTop Bottle Top Dispenser is a simple and easy-to-use solution for rapid and repetitive dispensing. Universal adapters make it simple to fit numerous bottles, and the engineering incorporates a variety of design options to assure precision and accuracy. Experience a design that was developed to provide un matched features and satisfaction.
Features
  • The updated design makes it simple to dismantle, clean, and reassemble the bottom unit.
  • Discharge tubes with recirculation valves prevent sample loss while also conserving valuable chemicals. (Recirculating Models Only)
  • A flexible telescopic filling tube that fits bottles with heights ranging from 170mm to 300mm.
  • Discharge tube with a 360-degree rotation for convenient dispensing from different bottle types.
  • Ergonomic design makes it easy to set and dispense even large volumes.
  • It is fully autoclavable with a safety locking function to prevent accidental spillage.
Properties
  • Unique Piston Design intuitive assembly allows for easy maintenannce
  • Springless Valve Design sprignless construction yields higher chemical resistance.
  • Volume Lock prevents accidental flow of reagents
  • Snap-Fit Mounting Easy handling and mobility of the discharge tube
  • Dual Graduation Markings users can clearly see and set the volume from multiple angles/dispenser positions
  • 4 Bottle Adapters for compatibility with varied bottle sizes
  • ISO 8655-5 Standards supplied with dedicated calibration report
Bottle Top Dispensers provide a unique solution to repetitive liquid handling challenges. The AccuTop Bottle Top Dispenser is specifically designed to handle a wide variety of chemicals and do so accurately, precisely, and conveniently. The AccuTop was engineered with a unique piston mechanism that protects the piston and cylinder without affecting calibration. Furthermore, the piston and cylinder can repeatedly dispense without the need for a spring. The springless valve design also enables smooth functioning and avoids chemical degradation that would otherwise affect the aspiration and dispensing.
Beyond just the engineered benefits, the AccuTop Bottle Top Dispenser is further designed for usability and convenience. The PTFE and PFA materials used in the construction of the dispenser offer further resistance against corrosive chemicals. Dual graduation marks along the dispenser handle improve readability and ensure that users can easily identify the volumes they are working with. The AccuTop Bottle Top Dispenser is also available in eco-friendly configurations. The re-circulating models allow users to prime the dispenser without needing to waste any reagent. These AccuTop Bottle Top Dispensers offer a full line of features that users need and the comfort that they enjoy!