Documentos clave
OM-130810
OMICRON™
OMICRON™ Glass Fiber Filters
Borosilicate Glass Fiber Binder Free Filter, 1.6 Micron, 8"X10" (203mm X 254mm), (100/Pk)
Sinónimos:
Glass Fiber, Filters, GF/A, Membrane
About This Item
PDF Icon
Shipped & Sold By Omicron Scientific
A Trusted Marketplace Seller
PDF Icon
Have a question?
General description
Product Description
OMICRON™ Glass Fiber filters are manufctured from pure Borosilicate Glass Fibers and are available in a wide range of nominal pore sizes. These filters have extremely low pressure drop, making them an excellent choice as a pre-filter for fluids that have high viscous and high dust holding capacity. Target applications include salt, sugar, sera, tissue lysates, protein solutions, and environmental samples, like soil and ground water, where there is an elevated level of suspended or un-dissloved particles. The production process of OMICRON™ Glass Fiber Filters has been validated with an accredited registered body to ISO 9001:2015 Quality System Standards.
Features and Benefits
  • Biologically inert
  • High dirt loading capacity
  • Available with Acrylic binders
  • Low resistance with high filtration efficiency
  • Hydrophilic material membrane
  • Low Extractable
Technical Applications
  • Stack sampling and Air pollution monitoring
  • Water pollution monitoring of effluents
  • Filtration of water, algae and bacteria cultures
  • Foodstuff analysis
  • Gravimetric determination of airborne particulates
(Cross Reference GF/A 1820-866)
Legal Information
OMICRON™ is a trademark of Omicron Scientific, Inc.