PCR96-AB-C-10
Novas Bio
Novas Bio 96-well PCR Plate
10 Plates, 0.2 ml 96-well PCR plate. Semi-skirted. Elevated skirt for ABI.
About This Item
material
polypropylene
size
96 wells
General description
- 96 well x 0.2 ml. Standard profile
- Single notch corner at A12
- Especially designed for ABI fast cyclers and sequencers:
- Raised wells for optimum seal and minimized evaporation
- Elevated skirt around plate provides optimum fit with ABI blocks
- Semi-skirted design allows for labeling
- Manufactured with medical-grade polypropylene
- Thin wall design for efficient heat transfer
- Exceptional clarity for maximum sample visibility
- Certified DNase, RNase, Human DNA free. Non-pyrogenic.
- Pairs perfectly with Novas Bio sequencing mat