Saltar al contenido
MilliporeSigma
Todas las fotos(1)

Documentos clave

MABS2145

Sigma-Aldrich

Anti-Polycystin-2 Antibody, clone YCE2

Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

antibody form

purified antibody

clone

YCE2
monoclonal

purified by

using protein G

species reactivity

human, mouse, mouse, rat, rat, human, mouse

concentration

(Please refer to lot specific datasheet.)

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

UniProt accession no.

General description

GST-tagged recombinant fragment corresponding to 68 amino acids from the C-terminal region, cytoplasmic domain.

Specificity

Clone YCE2 is a mouse monoclonal antibody that specifically detects Polycystin-2. It targets an epitope within 68 amino acids from the cytoplasmic domain in the C-terminal region.

Application

Epitope: C-terminus

Quality

Evaluated by Immunohistochemistry (Paraffin) in human fallopian tube tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution of this antibody detected Polycystin-2 in human fallopian tube tissue sections.

Physical properties

109.69 kDa calculated.

Physical form

Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Human, Mouse, Rat.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificados de análisis (COA)

Busque Certificados de análisis (COA) introduciendo el número de lote del producto. Los números de lote se encuentran en la etiqueta del producto después de las palabras «Lot» o «Batch»

¿Ya tiene este producto?

Encuentre la documentación para los productos que ha comprado recientemente en la Biblioteca de documentos.

Visite la Librería de documentos

Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.

Póngase en contacto con el Servicio técnico