Saltar al contenido
MilliporeSigma
Todas las fotos(1)

Documentos

MABS2026

Sigma-Aldrich

Anti-EphA2 Antibody, clone F2-27

clone F2-27, from mouse

Sinónimos:

Ephrin type-A receptor 2, EC: 2.7.10.1, Epithelial cell kinase, Tyrosine-protein kinase receptor ECK

Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

F2-27, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable
immunoprecipitation (IP): suitable
inhibition assay: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... EPHA2(1969)

General description

Ephrin type-A receptor 2 (UniProt: P29317; also known as EC: 2.7.10.1, Epithelial cell kinase, Tyrosine-protein kinase receptor ECK) is encoded by the EPHA2 (also known as ECK) gene (Gene ID: 1969) in human. Ephrin type-A receptor 2 is a homodimeric single-pass type I membrane protein that is expressed in brain and glioma tissue and in glioma cell lines. It is also highly expressed in tissues that contain a high proportion of epithelial cells. Its levels are shown to be up-regulated by UV irradiation via a TP53-independent, MAPK-dependent mechanism. Ephrin type-A receptor 2 serves as a receptor tyrosine kinase that binds promiscuously membrane-bound ephrin-A family ligands residing on adjacent cells, leading to contact-dependent bidirectional signaling into neighboring cells. It is involved in the regulation of migration, integrin-mediated adhesion, and in proliferation and differentiation of cells. Ephrin type-A receptor 2 is phosphorylated on tyrosine residues upon binding and activation by EFNA1. Phosphorylation on Tyr 588 and Tyr 594 is reported to be essential for binding VAV2 and VAV3 while phosphorylation on Tyr 735 and Tyr 930 are required for binding PI3-kinase p85 subunit. It is synthesized with a signal peptide (aa 1-23) that is cleaved off in the mature form. The mature form has an extracellular domain (aa 24-537), a short transmembrane domain (aa 538-558), and a cytoplasmic domain (aa 559-976). Two isoforms of Ephrin type-A receptor 2 have been described that are produced by alternative splicing. EphA2 is reported to be overexpressed in skin tumors murine models and its overexpression may represent a compensatory feedback mechanism during tumorigenesis. (Ref.: Guo, H., et al. (2006). Cancer Res. 66(14); 7050-7058).

Specificity

Clone F2-27 specifically detects Ephrin type -A receptor 2 in human.

Immunogen

KP-2, KP-3, SUIT-2, and MIAPaCa-2 cells.

Application

Anti-EphA2, clone F2-27, Cat. No. MABS2026, is a mouse monoclonal antibody that detects Ephrin type-A receptor 2 and has been tested for use in Flow Cytometry, Immunoprecipitation, and Inhibition Assay.
Inhibition Analysis: A representative lot inhibited cancer cell invasion in a three-dimensional spheroid formation assay. (Tanaka, T., et. al. (2017). Transl Oncol. 10(4):476-484).

Immunoprecipitation Analysis: A representative lot detected EphA2 in Immunoprecipitation applications (Tanaka, T., et. al. (2017). Transl Oncol. 10(4):476-484).

Flow Cytometry Analysis: A representative lot detected EphA2 in Flow Cytometry applications (Tanaka, T., et. al. (2017). Transl Oncol. 10(4):476-484).
Research Category
Signaling

Quality

Evaluated by Flow Cytometry in PC-3 cells.

Flow Cytometry Analysis: 1 µg of this antibody detected EphA2 in one million PC-3 cells.

Target description

108.27 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Herramienta de selección de productos.


Certificados de análisis (COA)

Busque Certificados de análisis (COA) introduciendo el número de lote del producto. Los números de lote se encuentran en la etiqueta del producto después de las palabras «Lot» o «Batch»

¿Ya tiene este producto?

Encuentre la documentación para los productos que ha comprado recientemente en la Biblioteca de documentos.

Visite la Librería de documentos

Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.

Póngase en contacto con el Servicio técnico