Saltar al contenido
MilliporeSigma
Todas las fotos(1)

Key Documents

HTS022M

Sigma-Aldrich

ChemiSCREEN MC3 Membrane Preparation

Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización


About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000
NACRES:
NA.45

biological source

human

Quality Level

recombinant

expressed in Chem-1 cells

form

liquid

manufacturer/tradename

ChemiScreen
Chemicon®

technique(s)

ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable

NCBI accession no.

UniProt accession no.

General description

Human MC3
The melanocortins, the peptides alpha-, beta- and gamma-melanocyte-stimulating hormone (MSH), bind to a family of five Gs-coupled GPCR receptors (MCRs) and play important roles in energy balance, reproductive function, pigmentation and inflammation (Gantz and Fong, 2003). MC3R is expressed in the CNS, including the hypothalamus, and appears to play a role in energy homeostasis as indicated by the observation that MC3R-null mice have increased subcutaneous fat, a loss of lean body mass, and reduced energy expenditure (Butler et al., 2000; Chen et al., 2000). Macrophages also express MC3R, which together with MC1R mediates the anti-inflammatory activity of melanocortins in animal models of peritonitis and myocardial infarct (Getting et al., 2003; 2004). Chemicon′s MC3R membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal HTS tools for screening of antagonists of MC3R interactions with melanocortins. The membrane preparations exhibit a Kd of 0.4 nM for [125I]-MSH. With 10.0 mg/well MC3R Membrane Prep and 0.25 nM [125I]-MSH, a greater than 7-fold signal-to-background ratio is obtained.

Application

Human MC3 GPCR membrane preparation for Radioligand binding Assays & GTPgammaS binding.
Radioligand binding assay, and GTPgammaS binding.

Biochem/physiol Actions

  • GPCR Class: A
  • Protein Target: MC3
  • Target Sub-Family: Melanocortin

Specifications

Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, a GF/C 96-well filter plate is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM HEPES, pH 7.4, 0.5% BSA. Binding reaction is transferred to the filter plate, and washed 9 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.Binding buffer: 25 mM Hepes, pH 7.0, 1 mM MgCl2, 1.5 mM CaCl2 2H2O, 100mM NaCl, 0.2% BSA, Protease Inhibitor, filter and stored at 4 °C.

Radioligand: [125I] MSH (Perkin Elmer# NEX352)

Wash Buffer: 25 mM Hepes, pH 7.0, 1 mM MgCl2, 1.5 mM CaCl2 2H2O, 100mM NaCl, 0.2% BSA, filtered and stored at 4°C. One vial contains enough membranes for at least 200 assays (units), where an unit is the amount of membrane that will yield greater than 4-fold signal:background with 125I-labeled MSH at 0.25 nM

Physical form

Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA no preservatives.Packaging method: Membranes protein were adjusted to 0.5 mg/ml in 1 ml packaging buffer, rapidly frozen, and stored at -80°C.

Storage and Stability

Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2


Certificados de análisis (COA)

Busque Certificados de análisis (COA) introduciendo el número de lote del producto. Los números de lote se encuentran en la etiqueta del producto después de las palabras «Lot» o «Batch»

¿Ya tiene este producto?

Encuentre la documentación para los productos que ha comprado recientemente en la Biblioteca de documentos.

Visite la Librería de documentos

Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.

Póngase en contacto con el Servicio técnico