Saltar al contenido
MilliporeSigma
Todas las fotos(1)

Key Documents

235400

Caspase-3 Substrate I, Colorimetric

Colorimetric substrate for caspase-3 (Km = 9.7 µM) and related cysteine proteases.

Sinónimos:

Caspase-3 Substrate I, Colorimetric, Ac-DEVD- pNA, Ac-DEVD-pNA

Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización


About This Item

Fórmula empírica (notación de Hill):
C26H34N6O13
Número de CAS:
Peso molecular:
638.58
UNSPSC Code:
12352200

assay

≥95% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

solubility

DMSO: 60 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

Colorimetric substrate for CPP32/Apopain/Yama (Km = 9.7 µM) and related cysteine proteases. Sequence is based on the P1-P4 tetrapeptide cleavage site of poly (ADP-ribose) polymerase (PARP) and includes Asp216. CPP32 is the protease responsible for the cleavage of PARP. Can be used with Pro-caspase-3 in in vitro assays to measure caspase-9 activity, since caspase-9 activates pro-caspase-3. Cleavage of the substrate can be monitored at 405 nm (ε = 9160 cm-1M-1).
Colorimetric substrate for caspase-3 (Km = 9.7 µM) and related cysteine proteases. Sequence is based on the P1-P4 tetrapeptide cleavage site of poly(ADP-ribose) polymerase (PARP) and includes Asp216. Caspase-3 is the protease responsible for the cleavage of PARP. Also a substrate for caspase-6, caspase-7, caspase-8, and caspase-10. Cleavage of pNA is monitored colorimetrically at ~405 nm.

Biochem/physiol Actions

Cell permeable: no
Km = 9.7 µM as colorimetric substrate for caspase-3
Primary Target
Caspas-3
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-Asp-Glu-Val-Asp-pNA

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Cardone, M.H., et al. 1998. Science 282, 1318.
Thornberry, N.A., and Lazebnik, Y. 1998. Science 281, 1312.
Nicholson, D.W., et al. 1995. Nature376, 37.
Lazebnik, Y.A., et al. 1994. Nature371, 346.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificados de análisis (COA)

Busque Certificados de análisis (COA) introduciendo el número de lote del producto. Los números de lote se encuentran en la etiqueta del producto después de las palabras «Lot» o «Batch»

¿Ya tiene este producto?

Encuentre la documentación para los productos que ha comprado recientemente en la Biblioteca de documentos.

Visite la Librería de documentos

Konrad A Szychowski et al.
Pesticide biochemistry and physiology, 168, 104638-104638 (2020-07-28)
Triclosan (TCS) is commonly used worldwide in a range of personal care and sanitizing products. A number of studies have revealed the presence of TCS in human tissues. It has recently been shown that TCS can interact with AhR in

Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.

Póngase en contacto con el Servicio técnico