Documentos
1.03835
Supelco
PTFE-ferrule for capillaries with 3.2 mm O.D., for HPLC bottle adapter S40
pack of 10
About This Item
UNSPSC Code:
41121516
packaging
pack of 10
storage temp.
2-30°C
General description
(pack of 10)
These PTFE-ferrules are spare parts for our HPLC bottle-adapters 1.03830.0001 and 1.03831.0001 to reduce the inner diameter of the tube connections to 3.2 mm. Ferrules are used before the fittings 1.03834.0001 are screwed in. Material: PTFE white
Storage Class
10-13 - German Storage Class 10 to 13