58819-05
MicroSolv
MicroSolv Mobile Phase Bottles for HPLC-Safety Coated
2 L Bottle, mobile phase reservoir, safety, coated glass, clear, screw top, GL45 threads. MicroSolv Brand. 1 EA.
Sinónimos:
Storage Bottles
About This Item
Productos recomendados
capacity
2 L
feature
with cap
General description
One of our Most Popular Items, this Clear, Plastic Coated, Borosilicate Glass Bottle has a 2L "Total Fill Capacity" and is perfect as an HPLC Mobile Phase Reservoir. The small opening, one liter Capacity and White Enamel Graduations on the outside of the Bottle help you determine the Solvent use and prevent running dry.
A Solid Screw Cap without a liner and a polypropylene Drip Ring are included.
The outside only of the Bottle is coated in a transparent, thick epoxy plastic and if this bottle is dropped and broken, the plastic will prevent a spill incident and prevent workers from getting cut with glass or splashed with solvent.