49225-80-10M
MicroSolv
MicroSolv ARE Brand PTFE Teflon Tubing
Tubing, PTFE, 0.011 inch (0.3 mm) ID, 0.031 inch ( 0.8 mm) OD, low pressure. 10 meter roll, ARE-Applied Research brand. 1 EA.
Sinónimos:
chromatography, PTFE Tubing, HPLC, Low Pressure
About This Item
O.D. × I.D. × L
0.8 mm x 0.3 mm x 10 m
General description
This thin-walled PTFE (Polytetrafluoroethylene) tubing is extruded from virgin PTFE. No additives are added to the original PTFE material.
This tubing is used widely in all low-pressure, laboratory applications including before and after an HPLC Pump such as with Mobile Phase Delivery and Solvent Waste Systems. Since PTFE tubing contains "micro-pores" in the tubing wall, which allows a quick air exchange, consideration for use with degassed and oxygen-sensitive solvents is needed.
Internal and Outer Dimensional Tolerance: +/- 0.10mm
Color: Natural, Translucent
Wall Thickness: 0.25 mm
Max Temperature: 260°C
Certificados de análisis (COA)
Busque Certificados de análisis (COA) introduciendo el número de lote del producto. Los números de lote se encuentran en la etiqueta del producto después de las palabras «Lot» o «Batch»
¿Ya tiene este producto?
Encuentre la documentación para los productos que ha comprado recientemente en la Biblioteca de documentos.