Saltar al contenido
MilliporeSigma
Todas las fotos(1)
Documentos clave
LABC-3525-350-000-9
Labcon
SuperClear 17x100mm Culture Tubes
Bags, 14mL, Quantity per Pack: 125, Clear, Polystyrene
Sinónimos:
Tube, 17x100mm, SuperClear
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
About This Item
PDF Icon
Shipped & Sold By Labcon
A Trusted Marketplace Seller
PDF Icon
Have a question?
El producto LABC-3525-350-000-9 no se vende actualmente en su país. Contact Labcon.
manufacturer/tradename
Labcon
General description
SuperClear® tubes are made from an ultra-clear medical grade resin and engineered with thick, durable walls for high speed centrifugation. They consistently outperform other brands and are designed to prevent breakage and leakage during centrifuge and freezing. Our leak-resistant flat caps keep your samples secure and our transparent, extra large writing panel is smudge-resistant and easy to see through. Tubes can also be autoclaved (122°C). Per Pack: 125 Per Case: 1000 Pack:Case Ratio: 8 Package Type: Bags Sterile: No Shelf Life (Months): 72 Shelf Life (Days): 2160 Volume: 14mL Color: Clear Material: Polystyrene
Legal Information
SuperClear® is a trademark of Labcon.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documentos section.
Si necesita más asistencia, póngase en contacto con Atención al cliente
¿Ya tiene este producto?
Visite la Librería de documentos