HS3301NG
Hausser Scientific
Hausser Scientific Counting Chamber - No Grid
Counting chamber, 100 micron cell depth, two counting areas, "V" groove loading, two cover slips
manufacturer/tradename
Hausser Scientific
Categorías relacionadas
General description
Counting chamber, 100 micron cell depth, two counting areas, "V" groove loading, two cover slips