137035-500
Globe Scientific
Globe Scientific Graduated Transfer Pipets
Transfer Pipet, 3.0mL, Small Bulb, Graduated to 1mL, 140mm, 500/Dispenser Box
About This Item
Productos recomendados
General description
Graduated transfer pipets are designed for measurements, these pipets feature molded-in graduation marks that will never fade or wash off.
Download a PDF copy of our transfer pipet brochure. A full size drawing of this pipette is featured along with technical details.
Material: Low Density Polyethylene (LDPE)
Packaging: Globe Scientific's non-sterile transfer pipets are packaged in a unique dispenser box. This convenient box is small and compact and ideal for bench-top use. The hinged front panel opens for quick access to the pipets and can be closed when not in use to keep the contents clean.
Certificados de análisis (COA)
Busque Certificados de análisis (COA) introduciendo el número de lote del producto. Los números de lote se encuentran en la etiqueta del producto después de las palabras «Lot» o «Batch»