MilliporeSigma
137035-500
Globe Scientific
Globe Scientific Graduated Transfer Pipets
Transfer Pipet, 3.0mL, Small Bulb, Graduated to 1mL, 140mm, 500/Dispenser Box
About This Item
PDF Icon
Shipped & Sold By Globe Scientific LLC
A Trusted Marketplace Seller
PDF Icon
Have a question?
General description
Graduated transfer pipets are designed for measurements, these pipets feature molded-in graduation marks that will never fade or wash off.
Download a PDF copy of our transfer pipet brochure. A full size drawing of this pipette is featured along with technical details.
Material: Low Density Polyethylene (LDPE)
Packaging: Globe Scientific's non-sterile transfer pipets are packaged in a unique dispenser box. This convenient box is small and compact and ideal for bench-top use. The hinged front panel opens for quick access to the pipets and can be closed when not in use to keep the contents clean.