MilliporeSigma
104A-PL612
Glas-Col®
Glas-Col® Temperature Control
Precision Full PID Control
Sinónimos:
Temperature Control Auto, PID Control
About This Item
PDF Icon
Shipped & Sold By Glas-Col
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
Glas-Col
General description
DigiTrol, auto tuning temperature control, digital readout, with universal input.
  • 1800W
  • 120 Volts
  • 0-750C
Legal Information
Glas-Col is a registered trademark of Glas-Col LLC