Key Documents
100A-O616
Glas-Col®
Glas-Col® Heating Mantle
2 Liter
Sinónimos:
heating mantle bottom, heating mantle
About This Item
PDF Icon
Shipped & Sold By Glas-Col
A Trusted Marketplace Seller
PDF Icon
Have a question?
AC/DC
115
feature
CSA approved
heater capacity
550W
manufacturer/tradename
Glas-Col
General description
Fabric mantle for 2 liter griffin beaker.
  • 550 Watts
  • 115 Volts
  • Max Temperature 400oC
  • CSA Approved
  • A controller is required for operating mantles
Legal Information
Glas-Col is a registered trademark of Glas-Col LLC