Documentos clave
252-1903-OEM
Foxx Life Sciences
Foxx Life Sciences EZwaste® HPLC Solvent Waste System, Secondary Container
HPLC Solvent Waste Safety Tray Secondary Container, for 20L-60L Carboys REV A, 1/EA
Seleccione un Tamaño
1 EA
$106.64
$106.64
Disponible para envío el07 de mayo de 2025Detalles
Seleccione un Tamaño
Cambiar Vistas
1 EA
$106.64
About This Item
PDF Icon
Shipped & Sold By Foxx Life Sciences
A Trusted Marketplace Seller
PDF Icon
Have a question?
$106.64
Disponible para envío el07 de mayo de 2025Detalles
fabricante / nombre comercial
Foxx Life Sciences
Descripción general
Foxx Life Sciences' Secondary Container is a perfect solution for preventing leakage and spillage from Foxx Life Sciences 1L, 2.5L, 4L, 5L, and 10L EZwaste® Solvent Waste System containers. Made from HDPE material, these containers are built to be tough and durable. Our Secondary Containers have the ability to hold at least 115% of the container's content.
Check our secondary containers' specifications to see if they fit your container.
Made to Order
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)