MTST1
Eisco Labs
Eisco Labs Metalware Set
4 Piece Set - Rectangular Retort Stand, Rod, Clamp & Ring Set
General description
- HIGH QUALITY || This laboratory set includes retort base made of corrosion resistant, powder coated steel, a stainless steel rod featuring a 10mm x 1.5mm metric thread, a support ring and a four-pronged cork-lined clamp
- STABILITY || The 8" long by 5" wide base weighs 1.75lbs, providing superb stability for experiments & setups. The included rod measures 23.6" tall and is 10mm in diameter
- TAPPED HOLE AND THREADED ROD || Base features a tapped hole, which fits the included rod, and can accommodate other rods with a 10mm x 1.5mm metric thread
- GREAT FOR CLASSROOM & HOME LABORATORIES || The durable and sturdy construction of this set makes it an excellent choice for science classrooms, household laboratories & basic professional laboratory setups