Saltar al contenido
MilliporeSigma
Todas las fotos(1)
Documentos clave
MTST1
Eisco Labs
Eisco Labs Metalware Set
4 Piece Set - Rectangular Retort Stand, Rod, Clamp & Ring Set
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
About This Item
PDF Icon
Shipped & Sold By Eisco Labs
A Trusted Marketplace Seller
PDF Icon
Have a question?
General description
  • HIGH QUALITY || This laboratory set includes retort base made of corrosion resistant, powder coated steel, a stainless steel rod featuring a 10mm x 1.5mm metric thread, a support ring and a four-pronged cork-lined clamp
  • STABILITY || The 8" long by 5" wide base weighs 1.75lbs, providing superb stability for experiments & setups. The included rod measures 23.6" tall and is 10mm in diameter
  • TAPPED HOLE AND THREADED ROD || Base features a tapped hole, which fits the included rod, and can accommodate other rods with a 10mm x 1.5mm metric thread
  • GREAT FOR CLASSROOM & HOME LABORATORIES || The durable and sturdy construction of this set makes it an excellent choice for science classrooms, household laboratories & basic professional laboratory setups
Certificados de análisis (COA)
Busque Certificados de análisis (COA) introduciendo el número de lote del producto. Los números de lote se encuentran en la etiqueta del producto después de las palabras «Lot» o «Batch»
¿Ya tiene este producto?
Visite la Librería de documentos