Key Documents
HTMST100V1
Eisco Labs
Eisco Labs Hot Plate w/ Magnetic Stirrer
Hot Plate w/ Magnetic Stirrer-110/120 V AC
Sinónimos:
Hot Plate, Hot Plate Stirrer
About This Item
PDF Icon
Shipped & Sold By Eisco Labs
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
Eisco Labs
General description
  • Surface temperature is adjustable up to 350°C (662°F)
  • Heating power output of 180 watts
  • Stirrer adjustable from 100 to about 2000 rpm
  • Plate size approx 4.75"x4.75"
  • Plate is Aluminum
This economical lab hotplate includes an integrated magnetic stirrer. The dual controls allow you to adjust the hotplate heating surface temperature and the stirring speed independently, which makes it ideal for light duty laboratory use. The hot plate heating surface temperature is adjustable up to 350°C (662°F) and reaches maximum temperature in a few minutes. The hot plate has a heating power output of 180 watts-enough to heat 500 ml of water to boiling in about 20 minutes at a low stirring speed. The magnetic stirrer is adjustable from 100 to about 2000 rpm and capable of stirring up to 2,000 ml liquid volume. Plate size 120 x 120 mm, plate is made of aluminium, power 180 W, Max Temp ~ 350°C, stirrer speed 100 ~ 2000 rpm, max. volume 2,000 ml.