CHWB1002
Eisco Labs
Eisco Labs LDPE Chemical Wash Bottle
Chemical LDPE Wash Bottle, No Label, 500ml - Wide Mouth
Sinónimos:
Wash Bottle, Wide Mouth, Wide Mouth Wash Bottle, Squeeze Bottle
About This Item
manufacturer/tradename
Eisco Labs
General description
- Unlabeled chemical wash bottle, 500ml capacity
- Wide Mouth for easy filling. Self venting design to eliminate leaks and dripping
- Easy to control squeeze for dispensing
- Performance Plastics by Eisco Labs || Superior quality low density polyethylene (LDPE) construction with long lasting strength. Responsive, with smooth bounce back that won't get stiff over time. Flex Ink won't fade, crack or peel.
Wash bottle made with Eisco Labs exclusive Performance Plastic. Made of low density polyethylene (LDPE) for superior quality, strength. Innovative Flex Ink won't fade, crack or peel.
No label design does offer a place to write contents. Wide mouth for easy refilling. Each bottle features a two piece construction, consisting of the bottle and leak proof cap with built in nozzle. Self venting design eliminates leaks and dripping. The low density polyethylene plastic is very responsive, with smooth bounce back that won't get stiff over time. Easy to control squeeze for dispensing.
500ml capacity.